missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble), Reagents

Catalog No. 01336737
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336737 10 g
01336736 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336737 Supplier Labchem Inc Supplier No. C611850010C1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.30
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[[4-(Dimethylamino)phenyl]diazenyl]benzoic Acid
IUPAC Name 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O

Specifications

Grade ACS
Packaging Glass Bottle
Specific Gravity 0.79
Color Brown to Red
Quantity 10 g
Physical Form Solid
Chemical Name or Material Methyl Red, ACS Reagent (Free Acid) (Acid Red 2 Not water soluble)