missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, ACS

Catalog No. AA3668222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3668222 100 g
AA3668214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3668222 Supplier Thermo Scientific Chemicals Supplier No. 03668222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol

pH indicator in pH range 4.2 - 6.3Methyl red is used as a pH indicator. It is red in pH less than 4.4 and yellow in pH over 6.2. It is used to study its degradation using Fenton's reagent. In microbiology, it is used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym C.I. 13020
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 179°C to 182°C
Beilstein 750102
Color Maroon to Red
Solubility Information Practically insoluble in water. Soluble in alcohol,acetic acidSoluble in ethanol. Insoluble in water.
Quantity 100 g
Physical Form Crystalline
Formula Weight 269.3
Grade ACS
Chemical Name or Material Methyl Red, For analysis ACS
Hazard Category H373
Precautionary Statement P260-P314-P501c
EINECSNumber 207-776-1
RTECSNumber DG8960000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only