missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Red, ACS
Description
pH indicator in pH range 4.2 - 6.3Methyl red is used as a pH indicator. It is red in pH less than 4.4 and yellow in pH over 6.2. It is used to study its degradation using Fenton's reagent. In microbiology, it is used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 179°C to 182°C |
| Beilstein | 750102 |
| Color | Maroon to Red |
| Solubility Information | Practically insoluble in water. Soluble in alcohol,acetic acidSoluble in ethanol. Insoluble in water. |
| Quantity | 25 g |
| Physical Form | Crystalline |
| Formula Weight | 269.3 |
| Grade | ACS |
| Chemical Name or Material | Methyl Red, For analysis ACS |
Safety and Handling
| Hazard Category | H373 |
| Precautionary Statement | P260-P314-P501c |
| EINECSNumber | 207-776-1 |
| RTECSNumber | DG8960000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only