missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red

CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol

Supplier:  Thermo Scientific Chemicals A1669036

 View more versions of this product

Catalog No. AAA1669036


Only null left

Description

Description

Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose

Solubility
Insoluble in water. Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Methyl Red
493-52-7
MFCD00002425
14,6119
Insoluble in water. Soluble in ethanol.
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
269.304
269.3
179°C to 182°C
C15H15N3O2
750102
C.I. 13020
CEQFOVLGLXCDCX-UHFFFAOYSA-N
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
10303
500 g

Chemical Identifiers

493-52-7
269.304
CEQFOVLGLXCDCX-UHFFFAOYSA-N
10303
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
C15H15N3O2
MFCD00002425
C.I. 13020
2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P260-P314-P501c

H373

EINECSNumber : 207-776-1

RTECSNumber : DG8960000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only