missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red (0.1% in ca. 95% Ethanol) [for Titration], TCI America™

Catalog No. M0067100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0067100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0067100ML Supplier TCI America Supplier No. M0067100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym methyl red, o-methyl red, c.i. acid red 2, acid red 2, cerven methylova, cerven kysela 2, 4-dimethylamino-2'-carboxylazobenzene, 2-carboxy-4'-dimethylamino azobenzene, 2-4-dimethylaminophenylazo benzoic acid, ci acid red 2
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Color Red
pH 4.2 - 6.2
Quantity 100 mL
UN Number 1170
Formula Weight 269.30
Physical Form Liquid
Chemical Name or Material Methyl Red (0.1% in ca. 95% Ethanol) [for Titration]
RTECSNumber DG8960000
TSCA Yes