missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(+)-lactate, 98%

Catalog No. AC414490250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414490250 25 g Glass bottle
AC414490050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414490250 Supplier Thermo Scientific Chemicals Supplier No. 414490250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17392-83-5 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17392-83-5
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004517
InChI Key LPEKGGXMPWTOCB-UHFFFAOYNA-N
Synonym methyl r-+-lactate, methyl d-lactate, r-methyl 2-hydroxypropanoate, methyl r-lactate, +-methyl d-lactate, methyl 2r-2-hydroxypropanoate, d-lactic acid methyl ester, methyl lactate, +, unii-45mz1t3tbv, r-methyl lactate
PubChem CID 637514
ChEBI CHEBI:74611
SMILES COC(=O)C(C)O

Specifications

Color Colorless
Density 1.0900g/mL
Boiling Point 144.0°C to 145.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4120 to 1.4140
Quantity 25 g
UN Number 3272
Specific Gravity 1.09
Specific Rotation Condition + 8.50 (20.00°C c=neat)
Specific Rotation + 8.50
Solubility Information Solubility in water: miscible,hydrolyses. Other solubilities: soluble in alcohol and ether,miscible with most common organic solvents
IUPAC Name methyl (2R)-2-hydroxypropanoate
Formula Weight 104.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl (R)-(+)-lactate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-420-6

RUO – Research Use Only