missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-3-Hydroxyisobutyrate 99.0+%, TCI America™

Catalog No. H07035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07035G 5 g
H070325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07035G Supplier TCI America Supplier No. H07035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72657-23-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00063450
InChI Key ATCCIZURPPEVIZ-SCSAIBSYSA-N
Synonym r-methyl 3-hydroxy-2-methylpropanoate, r---3-hydroxy-2-methylpropionic acid methyl ester, methyl r---3-hydroxy-2-methylpropionate, --methyl d-beta-hydroxyisobutyrate, methyl r---3-hydroxyisobutyrate, roche ester, methyl r-3-hydroxy-2-methylpropionate, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2r, methyl 2r-3-hydroxy-2-methylpropanoate, r---3-hydroxyisobutyric acid methyl ester
PubChem CID 5324733
IUPAC Name methyl (2R)-3-hydroxy-2-methylpropanoate
SMILES CC(CO)C(=O)OC

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 118.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (R)-(-)-3-Hydroxyisobutyrate