missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-3-Hydroxybutyrate 98.0+%, TCI America™

Catalog No. H070510ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H070510ML 10 mL
H070525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H070510ML Supplier TCI America Supplier No. H070510ML

Chemical Identifiers

CAS 3976-69-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00063289
InChI Key LDLDJEAVRNAEBW-SCSAIBSYSA-N
Synonym methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester
PubChem CID 2724279
IUPAC Name methyl (3R)-3-hydroxybutanoate
SMILES CC(CC(=O)OC)O

Specifications

Color Yellow
Boiling Point 75°C
Quantity 10 mL
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (R)-(-)-3-Hydroxybutyrate
EINECSNumber (2)-1430
RTECSNumber ET4700000
TSCA No