missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-3-hydroxybutyrate, 98%

Catalog No. AAL1415103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1415103 1 g
AAL1415106 5 g
AAL1415114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1415103 Supplier Thermo Scientific Chemicals Supplier No. L1415103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3976-69-0 | C5H10O3 | 118.132 g/mol

Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral β-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions, for use in the synthesis of chiral ß-lactams.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 3976-69-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00063289
InChI Key LDLDJEAVRNAEBW-SCSAIBSYSA-N
Synonym methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester
PubChem CID 2724279
IUPAC Name methyl (3R)-3-hydroxybutanoate
SMILES CC(CC(=O)OC)O

Specifications

Density 1.055
Boiling Point 56°C to 58°C (11 mmHg)
Flash Point 73°C (163°F)
Odor Pleasant
Refractive Index 1.422
Quantity 1 g
Beilstein 3648161
Solubility Information Slightly soluble in water.
Optical Rotation −48.5° (c=1.3 in Chloroform)
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Methyl (R)-(-)-3-hydroxybutyrate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H318-H227-H315
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 223-610-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only