missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pyruvate 97.0+%, TCI America™

Catalog No. P058025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P058025ML 25 mL
P0580500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P058025ML Supplier TCI America Supplier No. P058025ML

Chemical Identifiers

CAS 600-22-6
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008754
InChI Key CWKLZLBVOJRSOM-UHFFFAOYSA-N
Synonym methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester
PubChem CID 11748
ChEBI CHEBI:51850
IUPAC Name methyl 2-oxopropanoate
SMILES CC(=O)C(=O)OC

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 mL
UN Number 3272
Formula Weight 102.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Pyruvate
EINECSNumber (2)-1505
TSCA No