missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyruvate, 98%

Catalog No. p-7021574
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1396614 25 g
AAA1396622 100 g
AAA1396636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1396614 Supplier Thermo Scientific Chemicals Supplier No. A1396614
Only null left

CAS: 600-22-6 | C4H6O3 | 102.089 g/mol

Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.

Solubility
Soluble in chloroform, methanol, ether, alcohol. Slightly soluble in water.

Notes
Store away from oxidizing agent, heat.

Chemical Identifiers

CAS 600-22-6
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008754
InChI Key CWKLZLBVOJRSOM-UHFFFAOYSA-N
Synonym methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester
PubChem CID 11748
ChEBI CHEBI:51850
IUPAC Name methyl 2-oxopropanoate
SMILES CC(=O)C(=O)OC

Specifications

Density 1.123
Boiling Point 135°C to 137°C
Flash Point 39°C (102°F)
Assay Percent Range 98%
Refractive Index 1.404
Quantity 25 g
UN Number UN3272
Beilstein 1361953
Merck Index 14,8021
Solubility Information Soluble in chloroform,methanol,ether,alcohol. Slightly soluble in water.
Formula Weight 102.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl Pyruvate
Show More Show Less
Hazard Category H226-H317-H318-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 209-987-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only