missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyruvate, 94%

Catalog No. p-3779193
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127650250 25 g Glass bottle
AC127651000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC127650250 Supplier Thermo Scientific Chemicals Supplier No. 127650250
Only null left

CAS: 600-22-6 | C4H6O3 | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 600-22-6
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00008754
InChI Key CWKLZLBVOJRSOM-UHFFFAOYSA-N
Synonym methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester
PubChem CID 11748
ChEBI CHEBI:51850
IUPAC Name methyl 2-oxopropanoate
SMILES CC(=O)C(=O)OC

Specifications

Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 134.0°C to 137.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Refractive Index 1.4065
Linear Formula CH3COCO2CH3
Quantity 25 g
Beilstein 03, 616
Specific Gravity 1.13
Merck Index 15, 8135
Solubility Information Solubility in water: soluble.
Formula Weight 102.09
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Methyl pyruvate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-987-4

RUO – Research Use Only