missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyrrole-3-carboxylate, 97%

Catalog No. p-7036095
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5434003 1 g
AAH5434006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5434003 Supplier Thermo Scientific Chemicals Supplier No. H5434003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2703-17-5 | C6H7NO2 | 125.127 g/mol

Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2703-17-5
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00967074
InChI Key WLBNVSIQCFHAQB-UHFFFAOYSA-N
Synonym methyl pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylic acid, methyl ester, methyl 3-pyrrolecarboxylate, 3-methoxycarbonyl-1h-pyrrole, pubchem22507, acmc-1cmqn, methylpyrrole-3-carboxylate, 4,5-methoxy-carbonylpyrrole, methyl pyrrole 3-carboxylate
PubChem CID 12361759
IUPAC Name methyl 1H-pyrrole-3-carboxylate
SMILES COC(=O)C1=CNC=C1

Specifications

Melting Point 85°C to 89°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material Methyl pyrrole-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only