missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Pyridine-2-carboxylate 98.0+%, TCI America™
$96.94 - $583.44
Chemical Identifiers
| CAS | 7-6-2459 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.14 |
| MDL Number | MFCD00038038 |
| InChI Key | NMMIHXMBOZYNET-UHFFFAOYSA-N |
| Synonym | methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate |
| PubChem CID | 75569 |
| IUPAC Name | methyl pyridine-2-carboxylate |
| SMILES | COC(=O)C1=CC=CC=N1 |
Chemical Identifiers
| 7-6-2459 | |
| 137.14 | |
| NMMIHXMBOZYNET-UHFFFAOYSA-N | |
| 75569 | |
| COC(=O)C1=CC=CC=N1 |
| C7H7NO2 | |
| MFCD00038038 | |
| methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate | |
| methyl pyridine-2-carboxylate |
Specifications
| 7-6-2459 | |
| 233°C | |
| MFCD00038038 | |
| methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate | |
| COC(=O)C1=CC=CC=N1 | |
| 137.14 | |
| 137.14 | |
| Liquid |
| Yellow | |
| C7H7NO2 | |
| 25 g | |
| NMMIHXMBOZYNET-UHFFFAOYSA-N | |
| methyl pyridine-2-carboxylate | |
| 75569 | |
| ≥98.0% (GC) | |
| Methyl Pyridine-2-carboxylate |
Safety and Handling
TSCA : Yes