missing translation for 'onlineSavingsMsg'
Learn More

Methyl Purple Indicator, pH 4.8 to 5.4 Purple to Green, Certified, LabChem™

Catalog No. LC170707
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC170707 125 mL
LC170701 500 mL
LC170702 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC170707 Supplier LabChem Supplier No. LC170707

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7732-18-5 | 291.28 g/mol

Specifications

Chemical Name or Material Methyl Purple Indicator
Name Note pH 4.8 to 5.4 Purple to Green
CAS Max % 84.9, 15, 0.06, 0.04
CAS 67-63-0, 3486-30-4, 845-10-3
Identification Passes Test
Packaging Poly Bottle
Linear Formula C15H14N3O2Na
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Nitrogen oxides
Color Black/Green
Solubility Information Soluble in water
Quantity 125 mL
Flash Point 33°C
Physical Form Liquid
Molecular Weight (g/mol) 291.28
Formula Weight 291.28
Grade Certified
Show More Show Less