missing translation for 'onlineSavingsMsg'
Learn More

Methyl Propargyl Ether 98.0+%, TCI America™

Catalog No. p-7132411
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
M08605ML 5 mL
M086025ML 25 mL
2 options

Catalog No. M08605ML

Supplier: TCI America M08605ML

Only null left

Chemical Identifiers

CAS 627-41-8
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00008578
InChI Key YACFFSVYSPMSGS-UHFFFAOYSA-N
Synonym Methyl 2-Propynyl Ether
PubChem CID 69393
IUPAC Name 3-methoxyprop-1-yne
SMILES COCC#C

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 mL
UN Number 3271
Formula Weight 70.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Propargyl Ether