missing translation for 'onlineSavingsMsg'
Learn More

Methyl potassium malonate, 98%

Catalog No. AAL2042922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2042922 100 g
AAL2042914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2042922 Supplier Thermo Scientific Chemicals Supplier No. L2042922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acts as an inhibitor in protein glycosylation and advanced glycosylation | CAS: 38330-80-2 | C4H5KO4 | 156.178 g/mol

Methyl potassium malonate is a protein advanced glycosylation inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl potassium malonate is a protein advanced glycosylation inhibitor.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 38330-80-2
Molecular Formula C4H5KO4
Molecular Weight (g/mol) 156.178
MDL Number MFCD00014021
InChI Key WWTULTKUWBKVGV-UHFFFAOYSA-M
Synonym potassium 3-methoxy-3-oxopropanoate, methyl potassium malonate, potassium methyl malonate, propanedioic acid, monomethyl ester, potassium salt, potassium monomethyl malonate, monomethyl malonate potassium salt, hydrogen methyl malonate potassium salt, monomethylpotassiummalonate, acmc-209vp4, ksc226a9d
PubChem CID 2724687
IUPAC Name potassium;3-methoxy-3-oxopropanoate
SMILES COC(=O)CC(=O)[O-].[K+]

Specifications

Melting Point ∽190°C (decomposition)
Color White
Quantity 100 g
Solubility Information Soluble in methanol.
Formula Weight 156.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl potassium malonate, Hygroscopic
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-886-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only