missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pivalate 98.0+%, TCI America™

Catalog No. p-7134621
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P046225ML 25 mL
P0462500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P046225ML Supplier TCI America Supplier No. P046225ML
Only null left

Chemical Identifiers

CAS 598-98-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008843
InChI Key CNMFHDIDIMZHKY-UHFFFAOYSA-N
Synonym methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
PubChem CID 69027
IUPAC Name methyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC

Specifications

Color Colorless
Boiling Point 102°C
Quantity 25 mL
UN Number 3272
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Pivalate
EINECSNumber (2)-0776
TSCA Yes