missing translation for 'onlineSavingsMsg'
Learn More

Methyl picolinate, 98%

Catalog No. AC438140050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438140050 5 g
AC438140250 25 g
AC438141000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC438140050 Supplier Thermo Scientific Chemicals Supplier No. 438140050

CAS: 2459-07-6 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7-6-2459
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key NMMIHXMBOZYNET-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=CC=N1

Specifications

Melting Point 14°C to 16°C
Color White
Density 1.1370g/mL
Boiling Point 95°C (1.0 hPa)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.137
Synonym methyl isonicotinate, isonicotinic acid methyl ester, 4-pyridinecarboxylic acid, methyl ester, 4-picolinic acid methyl ester, 4-methoxycarbonylpyridine, methylisonicotinate, methyl 4-pyridinecarboxylate, 4-carbomethoxypyridine, isonicotinic acid, methyl ester, i-nicotinic acid, methyl ester
IUPAC Name methyl pyridine-4-carboxylate
PubChem CID 227085
Formula Weight 137.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl picolinate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 219-545-2

RUO – Research Use Only