missing translation for 'onlineSavingsMsg'
Learn More

Methyl picolinate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 438140250

Catalog No. AC438140250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl picolinate
1.1370g/mL
103°C
97.5% min. (GC)
25g
methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate
COC(=O)C1=CC=CC=N1
137.14
137.14
2459-07-6
95°C (1.0 hPa)
Authentic
C7H7NO2
1.137
NMMIHXMBOZYNET-UHFFFAOYSA-N
methyl pyridine-2-carboxylate
75569
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 219-545-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only