missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenylpropiolate, 98%

CAS: 4891-38-7 | C10H8O2 | 160.17 g/mol

$138.67 - $138.67

Chemical Identifiers

CAS 4891-38-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00041685
InChI Key JFGWPXKGINUNDH-UHFFFAOYSA-N
Synonym methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
PubChem CID 138378
IUPAC Name methyl 3-phenylprop-2-ynoate
SMILES COC(=O)C#CC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC334590050
SDS View Documents
Thermo Scientific Chemicals
334590050
5 g Glass bottle
Each for $138.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4891-38-7
160.17
JFGWPXKGINUNDH-UHFFFAOYSA-N
138378
COC(=O)C#CC1=CC=CC=C1
C10H8O2
MFCD00041685
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
methyl 3-phenylprop-2-ynoate

Specifications

4891-38-7
109°C to 110°C (2.0 mmHg)
97.5% min. (GC)
C10H8O2
MFCD00041685
methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
COC(=O)C#CC1=CC=CC=C1
160.17
160.17
Liquid After Melting
Colorless to Yellow
Authentic
Glass bottle
1.5620 to 1.564
5 g
JFGWPXKGINUNDH-UHFFFAOYSA-N
methyl 3-phenylprop-2-ynoate
138378
98%
Methyl phenylpropiolate, 98%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only