missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenylpropiolate, 97%

Catalog No. AAB2458706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2458706 5 g
AAB2458714 25 g
AAB2458722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2458706 Supplier Thermo Scientific Chemicals Supplier No. B2458706

CAS: 4891-38-7 | C10H8O2 | 160.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4891-38-7
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00041685
InChI Key JFGWPXKGINUNDH-UHFFFAOYSA-N
Synonym methyl phenylpropiolate, methyl 3-phenylpropiolate, phenylpropynoic acid methyl ester, 2-propynoic acid, 3-phenyl-, methyl ester, methyl phenylpropargylate, phenylpropiolic acid methyl ester, propiolic acid, 3-phenyl-, methyl ester, methylphenylpropalate, propiolic acid, phenyl-, methyl ester, methyl phenylpropynoate
PubChem CID 138378
IUPAC Name methyl 3-phenylprop-2-ynoate
SMILES COC(=O)C#CC1=CC=CC=C1

Specifications

Density 1.083
Boiling Point 109°C to 110°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.562
Linear Formula C6H5C≡CCO2CH3
Quantity 5 g
Beilstein 472813
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material Methyl phenylpropiolate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only