missing translation for 'onlineSavingsMsg'
Learn More

Methyl orotate, 95%

Catalog No. p-3779167
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208780250 25 g Glass bottle
AC208781000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC208780250 Supplier Thermo Scientific Chemicals Supplier No. 208780250
Only null left

CAS: 6153-44-2 | C6H6N2O4 | 170.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6153-44-2
Molecular Formula C6H6N2O4
Molecular Weight (g/mol) 170.12
MDL Number MFCD00010564
InChI Key UUTDWTOZAWFKFW-UHFFFAOYSA-N
Synonym methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate
PubChem CID 80257
IUPAC Name methyl 2,4-dioxo-1H-pyrimidine-6-carboxylate
SMILES COC(=O)C1=CC(=O)NC(=O)N1

Specifications

Melting Point 244°C
Color Brown or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Beilstein 25, 254
Merck Index 15, 6972
Solubility Information Solubility in water: .. Other solubilities: soluble in 1n nh4oh
Formula Weight 170.12
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Methyl orotate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens
EINECSNumber 228-171-9

RUO – Research Use Only