missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange-Xylene Cyanol, pH 2.9 to 4.6 Lavender to Green, Certified, LabChem™

Catalog No. LC170401
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC170401 500 mL
LC170402 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC170401 Supplier LabChem Supplier No. LC170401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 547-58-0 | 327.34 g/mol

Specifications

Chemical Name or Material Methyl Orange-Xylene Cyanol
Name Note pH 2.9 to 4.6 Lavender to Green
CAS 547-58-0, 4463-44-9, 547-58-0
CAS Max % 99.8, 0.1, 0.1
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Density 1g/mL
Color Gray/Green
Solubility Information Soluble in water
Quantity 500 mL
Physical Form Liquid
Formula Weight 327.34
Grade Certified
Show More Show Less