missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange-Xylene Cyanol, Indicator Solution, Spectrum™ Chemical
Description
- Changes color from purple (pH = 3.0) to green (pH = 4.5)
Specifications
Specifications
| Chemical Name or Material | Methyl Orange-Xylene Cyanol |
| CAS | 547-58-0, 2650-17-1, 7732-18-5 |
| Grade | Chemical Solution |
| Packaging | Amber Glass Bottle |
| Color | Gray/Green |
| Quantity | 500 mL |
| Physical Form | Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.