missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Solution R, EP Reagent 1054802, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Methyl Orange Solution |
| Name Note | Reagent |
| CAS | 7732-18-5, 64-17-5, 67-63-0, 67-56-1, 547-58-0 |
| CAS Min % | 84.0, 12.22, 0.64, 0.5, 0.08 |
| CAS Max % | 88.0, 12.71, 0.67, 1.5, 0.09 |
| Grade | Indicator |
| Packaging | Natural Poly Bottle |
| Color | Orange |
| Quantity | 500 mL |
| Physical Form | Liquid |