missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange, Modified, Certified, LabChem™

Catalog No. LC170201
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC170201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC170201 Supplier LabChem Supplier No. LC170201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified grade modified methyl orange indicator solution for laboratory use.

Specifications

Chemical Name or Material Methyl Orange
Name Note Modified
CAS 547-58-0, 4463-44-9, 547-58-0
CAS Max % 99.976, 0.014, 0.01
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Density 1g/mL
Color Gray/Green
pH 3.2 to 4.4
Solubility Information Soluble in water
Quantity 500 mL
Physical Form Liquid
Molecular Weight (g/mol) 327.34
Formula Weight 327.34
Grade Certified
Show More Show Less