missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Indicator, 0.1% (w/v), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Methyl Orange Indicator |
| CAS | 547-58-0, 7732-18-5 |
| Concentration | 0.1% (w/v) |
| Percent Purity | 0.1% W/V |
| Grade | Indicator |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Color | Orange |
| pH | 3 to 4 |
| Quantity | 120 mL |
| Show More |