missing translation for 'onlineSavingsMsg'
Learn More

Methyl octanoate, 99%, analytical standard for GC, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 460350050

 View more versions of this product

Catalog No. AC460350050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

111-11-5
158.24
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
CHEBI:87432
CCCCCCCC(=O)OC
C9H18O2
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
8091
methyl octanoate

Specifications

Methyl octanoate
0.8780g/mL
73°C
98.5% min. (GC)
1.4165 to 1.4185
5g
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
methyl octanoate
8091
158.24
111-11-5
79.0°C (12.0 mmHg)
Authentic
C9H18O2
CH3(CH2)6COOCH3
0.878
Solubility in water: insoluble.
CCCCCCCC(=O)OC
158.24
CHEBI:87432
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-835-

SDS
Documents

Documents

RUO – Research Use Only