missing translation for 'onlineSavingsMsg'
Learn More

Methyl o-Toluate 98.0+%, TCI America™

Catalog No. p-7136199
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T029625ML 25 mL
T0296500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T029625ML Supplier TCI America Supplier No. T029625ML
Only null left

Chemical Identifiers

CAS 89-71-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008428
InChI Key WVWZECQNFWFVFW-UHFFFAOYSA-N
Synonym methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate
PubChem CID 33094
IUPAC Name methyl 2-methylbenzoate
SMILES CC1=CC=CC=C1C(=O)OC

Specifications

Color Yellow
Boiling Point 213°C
Quantity 25 mL
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl o-Toluate
EINECSNumber (3)-1380
TSCA Yes