missing translation for 'onlineSavingsMsg'
Learn More

Methyl o-toluate, 99%

CAS: 89-71-4 | C9H10O2 | 150.177 g/mol

Supplier:  Thermo Scientific Chemicals A1569522

 View more versions of this product

Catalog No. AAA1569522

Only null left
Explore available promotions

Description

Description

It has been used in the preparation of 2-(azidomethyl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used in the preparation of 2-(azidomethyl)benzoic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
Specifications

Specifications

Methyl o-toluate
1.072
82°C (179°F)
1.519
100 g
methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate
WVWZECQNFWFVFW-UHFFFAOYSA-N
methyl 2-methylbenzoate
33094
99%
89-71-4
207°C to 208°C
C9H10O2
MFCD00008428
2041846
Not miscible or difficult to mix in water.
CC1=CC=CC=C1C(=O)OC
150.177
150.18

Chemical Identifiers

89-71-4
150.177
WVWZECQNFWFVFW-UHFFFAOYSA-N
33094
CC1=CC=CC=C1C(=O)OC
C9H10O2
MFCD00008428
methyl o-toluate, methyl orthotoluate, o-toluic acid, methyl ester, methyl o-methylbenzoate, methyl toluate, benzoic acid, 2-methyl-, methyl ester, 2-methylbenzoic acid methyl ester, o-toluic acid methyl ester, methyl-o-methyl benzoate, 2-methyl methylbenzoate
methyl 2-methylbenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 201-932-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only