missing translation for 'onlineSavingsMsg'
Learn More

Methyl Nonadecafluorodecanoate 97.0+%, TCI America™

Catalog No. M19165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19165G Supplier TCI America Supplier No. M19165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 307-79-9
Color Yellow
Boiling Point 193°C
Molecular Formula C11H3F19O2
Quantity 5 g
MDL Number MFCD00153211
Synonym methyl perfluorodecanoate, methyl nonadecafluorodecanoate, decanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro-, methyl ester, acmc-1cjo4, perfluorodecanoic acid methyl ester, nonadecafluorodecanoic acid methyl ester, nonadecafluorodecanoic acid methyl ester; perfluorodecanoic acid methyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid methyl ester, methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecakis fluoranyl decanoate
InChI Key QJFHNYDPNSFJMR-UHFFFAOYSA-N
SMILES COC(=O)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoate
Molecular Weight (g/mol) 528.113
PubChem CID 2733270
Formula Weight 528.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Nonadecafluorodecanoate
Show More Show Less