missing translation for 'onlineSavingsMsg'
Learn More

Méthyl N,N-Diméthylanthranilate 97,0+%, TCI America™

Numéro de catalogue. D067510G
Modifier l'affichage
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D067510G 10 g
D067525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D067510G Fournisseur TCI America Code fournisseur D067510G

Identifiants chimiques

CAS 10072-05-6
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00025858
InChI Key ZCNSBHAIPOWHJE-UHFFFAOYSA-N
Synonym methyl 2-dimethylamino benzoate, methyl n,n-dimethylanthranilate, unii-77lqp0681t, n,n-dimethylanthranilic acid methyl ester, benzoic acid, 2-dimethylamino-, methyl ester, acmc-1bs8u, anthranilic acid, n,n-dimethyl-, methyl ester, methyl dimethyl anthranilate, methyln,n-dimethylanthranilate, methyl 2-dimethylamino-benzoate
PubChem CID 82336
IUPAC Name Méthyle 2-(diméthylamino)benzoate
SMILES CN(C)C1=CC=CC=C1C(=O)OC

Spécifications

Color Jaune
Boiling Point 131°C
Quantity 10 g
Formula Weight 179.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquide
Chemical Name or Material Methyl N,N-Dimethylanthranilate