missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl N-4-[2-(5-chloro-2-methoxybenzamido)ethyl]benzenesulfonylcarbamate, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 21165-77-5 |
| Flash Point | Not Applicable |
| Molecular Formula | C18H19ClN2O6S |
| Linear Formula | C18H19ClN2O6S |
| Quantity | 25 mg |
| Synonym | Methyl {{4-{2-[(5-chloro-2-methoxybenzoyl)amino]ethyl}phenyl}sulfonyl}carbamate |
| Molecular Weight (g/mol) | 426.87 |
| Formula Weight | 426.87 |
| Grade | BP |
| Show More |