missing translation for 'onlineSavingsMsg'
Learn More

Methyl (Methylthio)acetate 99.0+%, TCI America™

Catalog No. M22965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22965G 5 g
M229625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22965G Supplier TCI America Supplier No. M22965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16630-66-3
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00039809
InChI Key HZYCAKGEXXKCDM-UHFFFAOYSA-N
Synonym methyl methylthio acetate, methyl 2-methylthio acetate, methyl 2-methylsulfanyl acetate, acetic acid, methylthio-, methyl ester, methyl methylithio acetate, unii-gn58c28ml9, methyl methylthioacetate, acmc-1c4wc, methyl 3-methylthio acetate
PubChem CID 85522
IUPAC Name methyl 2-methylsulfanylacetate
SMILES COC(=O)CSC

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
UN Number 3272
Formula Weight 120.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (Methylthio)acetate
TSCA No
Recommended Storage Refrigerator