missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methacrylate Polymer, TCI America™

Catalog No. M008825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M008825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M008825G Supplier TCI America Supplier No. M008825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9011-14-7
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134349
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
Synonym Methacrylic Acid Methyl Ester Polymer, Poly(methyl Methacrylate), PMMA
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
SMILES COC(=O)C(C)(-*)C-*

Specifications

Color White
Physical Form Crystal-Powder at 20°C
Quantity 25 g
Chemical Name or Material Methyl Methacrylate Polymer
EINECSNumber (6)-0524
RTECSNumber TR0400000
TSCA Yes