missing translation for 'onlineSavingsMsg'
Learn More

Methyl Mercaptan Sodium Salt (ca. 15% in Water), TCI America™

Catalog No. M0096500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0096500G 500 g
M009625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0096500G Supplier TCI America Supplier No. M0096500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.08
MDL Number MFCD00174316
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
Synonym sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium
PubChem CID 4378561
IUPAC Name sodium;methanethiolate
SMILES C[S-].[Na+]

Specifications

Color Yellow
Physical Form Liquid
Quantity 500 g
UN Number 2920
Formula Weight 70.08
Chemical Name or Material Methyl Mercaptan Sodium Salt (ca. 15% in Water)
EINECSNumber (2)-2446
TSCA Yes