missing translation for 'onlineSavingsMsg'
Learn More

Methyl Malonyl Chloride 97.0+%, TCI America™

Catalog No. M23155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23155G 5 g
M231525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23155G Supplier TCI America Supplier No. M23155G

Chemical Identifiers

CAS 37517-81-0
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00013657
InChI Key UTBCRHAMJFMIIR-UHFFFAOYSA-N
Synonym methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate
PubChem CID 123460
IUPAC Name methyl 3-chloro-3-oxopropanoate
SMILES COC(=O)CC(=O)Cl

Specifications

Color Yellow
Boiling Point 59°C
Quantity 5 g
UN Number 3265
Formula Weight 136.53
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl Malonyl Chloride
TSCA No
Recommended Storage Refrigerator