missing translation for 'onlineSavingsMsg'
Learn More

Methyl malonamate, 98%

Catalog No. p-7028170
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2532614 25 g
AAB2532622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2532614 Supplier Thermo Scientific Chemicals Supplier No. B2532614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51513-29-2 | C4H7NO3 | 117.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51513-29-2
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.10
MDL Number MFCD00674533
InChI Key LSNSJCKGQREPDW-UHFFFAOYSA-N
Synonym methyl malonamate, methyl malonate monoamide, methyl 2-carbamoylacetate, propanoic acid, 3-amino-3-oxo-, methyl ester, acmc-20akfl, methyl carbamoylacetate, methyl-malonate monoamide, malonamic acid methyl ester, malonamidic acid methyl ester, methyl 3-amino-3-oxo-propanoate
PubChem CID 3595299
IUPAC Name methyl 3-amino-3-oxopropanoate
SMILES COC(=O)CC(N)=O

Specifications

Melting Point 38°C to 40°C
Quantity 25 g
Formula Weight 117.11
Percent Purity 98%
Chemical Name or Material Methyl malonamate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only