missing translation for 'onlineSavingsMsg'
Learn More

Methyl m-Toluate 98.0+%, TCI America™

Catalog No. T1258500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1258500ML 500 mL
T125825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1258500ML Supplier TCI America Supplier No. T1258500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-36-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008436
InChI Key CPXCDEMFNPKOEF-UHFFFAOYSA-N
Synonym methyl m-toluate, methyl 3-toluate, benzoic acid, 3-methyl-, methyl ester, methyl m-methylbenzoate, meta-toluic acid, methyl ester, 3-methylbenzoic acid methyl ester, methyl-m-methyl benzoate, m-toluic acid, methyl ester, methyl3-methylbenzoate, m-toluic acid methyl ester
PubChem CID 7435
IUPAC Name methyl 3-methylbenzoate
SMILES CC1=CC=CC(=C1)C(=O)OC

Specifications

Color Yellow
Boiling Point 115°C
Quantity 500 mL
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material Methyl m-Toluate
EINECSNumber (3)-1380
TSCA Yes