missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl m-toluate, 98%
Description
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Density | 1.062 |
| Boiling Point | 113°C (27 mmHg) |
| Flash Point | 95°C (203°F) |
| Refractive Index | 1.516 |
| Quantity | 50 g |
| Beilstein | 2043574 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 150.18 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl m-toluate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-753-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only