missing translation for 'onlineSavingsMsg'
Learn More

Methyl m-toluate, 98%

Catalog No. AAA1089330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1089330 250 g
AAA1089318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1089330 Supplier Thermo Scientific Chemicals Supplier No. A1089330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-36-5 | C9H10O2 | 150.177 g/mol

Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 99-36-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008436
InChI Key CPXCDEMFNPKOEF-UHFFFAOYSA-N
Synonym methyl m-toluate, methyl 3-toluate, benzoic acid, 3-methyl-, methyl ester, methyl m-methylbenzoate, meta-toluic acid, methyl ester, 3-methylbenzoic acid methyl ester, methyl-m-methyl benzoate, m-toluic acid, methyl ester, methyl3-methylbenzoate, m-toluic acid methyl ester
PubChem CID 7435
IUPAC Name methyl 3-methylbenzoate
SMILES CC1=CC=CC(=C1)C(=O)OC

Specifications

Density 1.062
Boiling Point 113°C (27 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.516
Quantity 250 g
Beilstein 2043574
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material Methyl m-toluate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-753-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only