missing translation for 'onlineSavingsMsg'
Learn More

Methyl Lactate 98.0+%, TCI America™

Catalog No. L000425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L000425ML 25 mL
L0004500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L000425ML Supplier TCI America Supplier No. L000425ML

Chemical Identifiers

CAS 547-64-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00066367
InChI Key LPEKGGXMPWTOCB-UHFFFAOYSA-N
Synonym methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate
PubChem CID 11040
ChEBI CHEBI:83221
IUPAC Name methyl 2-hydroxypropanoate
SMILES CC(C(=O)OC)O

Specifications

Color Colorless
Boiling Point 144°C
Quantity 25 mL
UN Number 3272
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Lactate
EINECSNumber (2)-1370
RTECSNumber OD5670000
TSCA Yes