missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl L-(+)-Mandelate 98.0+%, TCI America™
$35.41 - $42.87
Chemical Identifiers
| CAS | 21210-43-5 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00064246 |
| InChI Key | ITATYELQCJRCCK-UHFFFAOYNA-N |
| Synonym | methyl s-+-mandelate, s-+-methyl mandelate, s-methyl 2-hydroxy-2-phenylacetate, methyl 2s-2-hydroxy-2-phenylacetate, s-methyl mandelate, l-+-mandelic acid methyl ester, +-methyl l-mandelate, methyl l-+-mandelate, methyl hydroxy phenyl acetate #, methyl s-mandelate |
| PubChem CID | 643570 |
| IUPAC Name | methyl 2-hydroxy-2-phenylacetate |
| SMILES | COC(=O)C(O)C1=CC=CC=C1 |
Chemical Identifiers
| 21210-43-5 | |
| 166.18 | |
| ITATYELQCJRCCK-UHFFFAOYNA-N | |
| 643570 | |
| COC(=O)C(O)C1=CC=CC=C1 |
| C9H10O3 | |
| MFCD00064246 | |
| methyl s-+-mandelate, s-+-methyl mandelate, s-methyl 2-hydroxy-2-phenylacetate, methyl 2s-2-hydroxy-2-phenylacetate, s-methyl mandelate, l-+-mandelic acid methyl ester, +-methyl l-mandelate, methyl l-+-mandelate, methyl hydroxy phenyl acetate #, methyl s-mandelate | |
| methyl 2-hydroxy-2-phenylacetate |
Specifications
| 21210-43-5 | |
| White | |
| C9H10O3 | |
| 1 g | |
| ITATYELQCJRCCK-UHFFFAOYNA-N | |
| methyl 2-hydroxy-2-phenylacetate | |
| 643570 | |
| ≥98.0% (GC) | |
| Methyl L-(+)-Mandelate |
| 55°C | |
| 138°C | |
| MFCD00064246 | |
| methyl s-+-mandelate, s-+-methyl mandelate, s-methyl 2-hydroxy-2-phenylacetate, methyl 2s-2-hydroxy-2-phenylacetate, s-methyl mandelate, l-+-mandelic acid methyl ester, +-methyl l-mandelate, methyl l-+-mandelate, methyl hydroxy phenyl acetate #, methyl s-mandelate | |
| COC(=O)C(O)C1=CC=CC=C1 | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No