missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isonicotinate 99.0+%, TCI America™

Catalog No. p-7131230
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
I014125G 25 g
I0141100G 100 g
2 options

Catalog No. I014125G

Supplier: TCI America I014125G

Only null left

Chemical Identifiers

CAS 9-8-2459
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006427
InChI Key OLXYLDUSSBULGU-UHFFFAOYSA-N
Synonym methyl isonicotinate, isonicotinic acid methyl ester, 4-pyridinecarboxylic acid, methyl ester, 4-picolinic acid methyl ester, 4-methoxycarbonylpyridine, methylisonicotinate, methyl 4-pyridinecarboxylate, 4-carbomethoxypyridine, isonicotinic acid, methyl ester, i-nicotinic acid, methyl ester
PubChem CID 227085
IUPAC Name methyl pyridine-4-carboxylate
SMILES COC(=O)C1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 209°C
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Methyl Isonicotinate
EINECSNumber (5)-0733
TSCA Yes
Recommended Storage Refrigerator