missing translation for 'onlineSavingsMsg'
Learn More

Methyl isonicotinate, 98%

CAS: 2459-09-8 | C7H7NO2 | 137.14 g/mol

$137.65 - $137.65

Chemical Identifiers

CAS 9-8-2459
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006427
InChI Key OLXYLDUSSBULGU-UHFFFAOYSA-N
SMILES COC(=O)C1=CC=NC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127061000
SDS View Documents
Thermo Scientific Chemicals
127061000
100 mL Glass bottle
Each for $137.65
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9-8-2459
137.14
OLXYLDUSSBULGU-UHFFFAOYSA-N
C7H7NO2
MFCD00006427
COC(=O)C1=CC=NC=C1

Specifications

9-8-2459
Brown to Orange
207.0°C to 209.0°C
Authentic
Glass bottle
1.5120 to 1.5140
100 mL
1.161
trimethyl trimellitate, trimethyl 1,2,4-benzenetricarboxylate, methyl trimellitate, trimellitic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, trimethyl ester, unii-16o7dt5n66, 1,2,4-benzenetricarboxylic acid trimethyl ester, 1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester, 1,2,4-trimethyl benzene-1,2,4-tricarboxylate, acmc-1ckkm
COC(=O)C1=CC=NC=C1
137.14
137.14
Liquid
8.5°C
1.1610g/mL
82°C
97.5% min. (GC)
C7H7NO2
MFCD00006427
22, 46
10, 5034
OLXYLDUSSBULGU-UHFFFAOYSA-N
trimethyl benzene-1,2,4-tricarboxylate
17159
98%
Methyl isonicotinate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 219-546-8

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only