missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isobutyrate 99.0+%, TCI America™

Catalog No. p-7131217
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
I010625ML 25 mL
I0106500ML 500 mL
2 options

Catalog No. I010625ML

Supplier: TCI America I010625ML

Only null left

Chemical Identifiers

CAS 547-63-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008914
InChI Key BHIWKHZACMWKOJ-UHFFFAOYSA-N
Synonym methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
PubChem CID 11039
ChEBI CHEBI:73689
IUPAC Name methyl 2-methylpropanoate
SMILES CC(C)C(=O)OC

Specifications

Melting Point -85°C
Color Colorless
Boiling Point 92°C
Quantity 25 mL
UN Number 1237
Formula Weight 102.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Isobutyrate
EINECSNumber (2)-0779
RTECSNumber NQ5425000
TSCA Yes