missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isobutyrate 99.0+%, TCI America™

Catalog No. I010625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I010625ML 25 mL
I0106500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I010625ML Supplier TCI America Supplier No. I010625ML

Chemical Identifiers

CAS 547-63-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008914
InChI Key BHIWKHZACMWKOJ-UHFFFAOYSA-N
Synonym methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
PubChem CID 11039
ChEBI CHEBI:73689
IUPAC Name methyl 2-methylpropanoate
SMILES CC(C)C(=O)OC

Specifications

Melting Point -85°C
Color Colorless
Boiling Point 92°C
Quantity 25 mL
UN Number 1237
Formula Weight 102.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Isobutyrate
EINECSNumber (2)-0779
RTECSNumber NQ5425000
TSCA Yes