Learn More
Methyl isobutyrate, 98%
CAS: 547-63-7 | C5H10O2 | 102.133 g/mol
Supplier: Thermo Scientific Chemicals A14070AE
Description
Methyl isobutyrate is used as a flavoring agent. It was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl isobutyrate is used as a flavoring agent. It was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| Methyl isobutyrate | |
| -85°C | |
| 90°C to 92°C | |
| Pleasant | |
| 1.384 | |
| 100 mL | |
| 1740720 | |
| methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne | |
| BHIWKHZACMWKOJ-UHFFFAOYSA-N | |
| methyl 2-methylpropanoate | |
| 11039 | |
| 102.13 |
| 547-63-7 | |
| 0.891 | |
| 3°C (37°F) | |
| C5H10O2 | |
| MFCD00008914 | |
| UN1237 | |
| 14,6088 | |
| Slightly soluble in water. | |
| CC(C)C(=O)OC | |
| 102.133 | |
| CHEBI:73689 | |
| 98% |
Chemical Identifiers
| 547-63-7 | |
| 102.133 | |
| BHIWKHZACMWKOJ-UHFFFAOYSA-N | |
| 11039 | |
| methyl 2-methylpropanoate |
| C5H10O2 | |
| MFCD00008914 | |
| methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne | |
| CHEBI:73689 | |
| CC(C)C(=O)OC |
Safety and Handling
GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL BUTYRATE
EINECSNumber : 208-929-5
RTECSNumber : NQ5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only