missing translation for 'onlineSavingsMsg'
Learn More

Methyl isobutyrate, 98%

Catalog No. AAA14070AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14070AP 500 mL
AAA14070AE 100 mL
AAA140700F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA14070AP Supplier Thermo Scientific Chemicals Supplier No. A14070AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 547-63-7 | C5H10O2 | 102.133 g/mol

Methyl isobutyrate is used as a flavoring agent. It was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl isobutyrate is used as a flavoring agent. It was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 547-63-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008914
InChI Key BHIWKHZACMWKOJ-UHFFFAOYSA-N
Synonym methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
PubChem CID 11039
ChEBI CHEBI:73689
IUPAC Name methyl 2-methylpropanoate
SMILES CC(C)C(=O)OC

Specifications

Melting Point -85°C
Density 0.891
Boiling Point 90°C to 92°C
Flash Point 3°C (37°F)
Odor Pleasant
Refractive Index 1.384
Quantity 500 mL
UN Number UN1237
Beilstein 1740720
Merck Index 14,6088
Solubility Information Slightly soluble in water.
Formula Weight 102.13
Percent Purity 98%
Chemical Name or Material Methyl isobutyrate
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL BUTYRATE
EINECSNumber 208-929-5
RTECSNumber NQ5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only